Learn More
Bel-Art™ SP Scienceware™ Swing Jaw™ Tubing Clamps
Provide nonslip closures for flexible tubing
Specifications
Type Tubing Clamp
Material Zinc-plated Steel
Jaw Type Swing
View More Specs
Description
Description
  • Made of zinc-plated steel
  • Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
  • Removable from tubing without disconnecting apparatus
  • Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
  • Strong enough to use with heavy wall vacuum lines
Specifications
Specifications
Tubing Clamp
Zinc-plated Steel
Swing