missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Stainless-Steel Spoons

Double-ended spoons offer two bowl sizes for convenient sample handling

Brand:  Bel-Art™ H36729-0030

 View more versions of this product

Product Code. 15580122

  • 110.00 € / Pack of 2
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Double-ended spoons offer two bowl sizes for convenient sample handling

  • Polished 304 stainless-steel construction
  • Solid, rounded shaft for easy handling

Specifications

Specifications

Spoon
Autoclavable
2, 8 mL
Silver
11.81 in.
30 cm
1.5 in.
Stainless Steel
Double end with 2mL small spoon at one end and 8mL large spoon at other
Reusable
11.81 x 1.5 in. (30 x 3.81 cm)
9.25 in.
23.49 cm
3.81 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.