missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Economy Dippers

For convenient sampling from tanks, vats, streams, ponds, lakes

Brand:  Bel-Art™ F36784-0016

 View more versions of this product

Product Code. 16669343

  • 82.50 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lightweight and portable
  • Thread on 500mL (16 oz.) polypropylene cup can be exchanged to reduce cross contamination
  • Two pouring spouts on cup allow left or right pouring
  • High-density polyethylene plastic handles are strong honeycomb tubes without inserts and are ribbed for sure grip
  • Handle end has a hook for hanging storage

Specifications

Specifications

Dipper
Autoclavable
500 mL
3 ft.
HDPE
16 oz.
For sampling from tanks, ponds, lakes
0.9 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers