Learn More
BDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15417-1-AP-20UL
 View more versions of this product
Product Code. 16894574
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This gene encodes a member of the short-chain dehydrogenase/reductase gene family. The encoded protein forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific requirement for phosphatidylcholine for optimal enzymatic activity. The encoded protein catalyzes the interconversion of acetoacetate and -3-hydroxybutyrate, the two major ketone bodies produced during fatty acid catabolism. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
Specifications
BDH1
Polyclonal
Unconjugated
BDH1
BDH, BDH1, SDR9C1
Rabbit
Antigen Affinity Chromatography
RUO
117099, 622, 71911
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P29147, Q02338, Q80XN0
BDH1
BDH1 Fusion Protein Ag7705
20 μL
Primary
Rat, Mouse, Human