missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Shigella Poly Antisera
Used in serologic identification of Shigella
Brand: BD 228381
This item is not returnable.
View return policy
Description
- Available in 3.0mL packaged vials
Specifications
2°C to 8°C | |
Antiserum | |
3 mL |
Liquid | |
Used for serologic identification of Shigella |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Vial Alkalescens-Dispar Antiserum Poly
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide