missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ HYcheck™ Contact Slides: Total Count

For control of cutting oils and process water. BD Difco™ HYcheck™ Contact Slides: Total Count are designed for environmental testing.

Brand:  BD 290005

Product Code. 11743563

  • 55.85 € / Pack of 10
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed for environmental testing
  • Double sided with Plate Count Agar and Plate Count Agar with TTC
  • Hinged paddle bends for easy sampling

Order Info

Ten units per pack.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light Amber
Solid
Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L
Slide
2°C to 8°C
United States
To assess microbiological contamination of surfaces or fluids
10 U
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers