Learn More
BD BBL™ Mueller Hinton II Agar, Cation-Adjusted
Recommended for antimicrobial susceptibility testing of common, rapidly growing bacteria. BD™ BBL™ Mueller Hinton II Agar is used in disc diffusion testing by the Bauer-Kirby agar diffusion method, as standardized by the Clinical and Laboratory Standards Institute (CLSI).
249.00 €
Specifications
Certifications/Compliance | BAM, BS12, CCAM, CLSI, CMPH2, ISO, MCM9 |
---|---|
Color | Light beige |
Form | Dehydrated (free-flowing, homogenous) |
Media Type | Mueller Hinton II Agar |
Product Type | Agar |
Description
Mueller Hinton Agar was originally developed for the cultivation of pathogenic Neisseria. It is recommended for antimicrobial disc diffusion susceptibility testing of common, rapidly growing aerobic pathogens. Unsupplemented Mueller Hinton agar is not adequate for more fastidious organisms such as S. pneumonia.
- Tested according to, and meets the acceptance limits of, the current M6 protocol published by the CLSI.
- Complies with requirements of the World Health Organization and is specified in the FDA Bacteriological Analytical Manual for food testing.
- Manufactured to contain low levels of thymine and thymidine, with controlled levels of calcium and magnesium.
- Displays a cultural response to: Enterococcus faecalis, Escherichia coli, Pseudomonas aeruginosa and Staphylococcus aureus.
Specifications
BAM, BS12, CCAM, CLSI, CMPH2, ISO, MCM9 | |
Dehydrated (free-flowing, homogenous) | |
Agar | |
Antimicrobial susceptibility testing of microorganisms by Agar diffusion method |
Light beige | |
Mueller Hinton II Agar | |
Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L |
Safety and Handling
missing translation for 'shelfLife' : 1460 Days
Your input is important to us. Please complete this form to provide feedback related to the content on this product.