missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bathocuproinedisulfonic acid, disodium salt hydrate, 97%, Thermo Scientific Chemicals
Bathocuproinedisulfonic acid, disodium salt hydrate, 97%, 2NaO3S.xH2O, CAS Number 98645-85-3
108.54 € - 1119.57 €
Chemical Identifiers
CAS | 98645-85-3 |
---|---|
Molecular Formula | C26H18N2Na2O6S2 |
Molecular Weight (g/mol) | 564.54 |
MDL Number | MFCD00149974 |
InChI Key | RNGKZLRAVYPLJC-UHFFFAOYSA-L |
Synonym | disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate |
PubChem CID | 15678335 |
IUPAC Name | disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate |
SMILES | [Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10684832
|
Thermo Scientific Acros
164060010 |
1 g | Glass bottle |
108.54 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10063993
|
Thermo Scientific Acros
164060050 |
5 g | Glass bottle |
255.27 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11956611
|
Thermo Scientific Acros
164060250 |
25 g | Glass bottle |
1119.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98645-85-3 | |
564.54 | |
RNGKZLRAVYPLJC-UHFFFAOYSA-L | |
15678335 | |
[Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O |
C26H18N2Na2O6S2 | |
MFCD00149974 | |
disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate |
Specifications
98645-85-3 | |
disulfonate) (HPLC) | |
MFCD00149974 | |
Solubility in water: soluble in water | |
[Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O | |
564.54 | |
564.54 | |
Bathocuproinedisulfonic acid, disodium salt hydrate |
300.0°C | |
C26H18N2Na2O6S2 | |
disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
RNGKZLRAVYPLJC-UHFFFAOYSA-L | |
disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
15678335 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 258-111-7
RUO – Research Use Only