Learn More
Memmert™ Basic Series Water Baths
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
Brand:  Memmert™ WNB10
 View more versions of this product
Product Code. 10329342
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Specifications
Microprocessor PID
CE, VDE, GS
For research laboratories, food and beverage, temperature control
WNB 10
±0.1°C
350 mm
210 mm
140 mm
17 kg
230 V
50/60 Hz
10 L
Digital LED
Stainless Steel
95°C
578 mm
356 mm
238 mm
350 mm
13 kg
1.2 kW
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Basic Series Water Baths > Capacity: 10L