missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Basic Fuchsin, Electrophoresis Reagent
20.38 € - 211.00 €
Specifications
CAS | 632-99-5 |
---|---|
Content And Storage | Ambient temperatures |
Format | Crystalling powder |
Formula Weight | 337.86 |
Melting Point | 268° to 270°C (dec.) |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
632-99-5 | |
Crystalling powder | |
268° to 270°C (dec.) | |
Plastic bottle |
Ambient temperatures | |
337.86 | |
C20H20ClN3 | |
C.I. 42510,Fuchsin basic |