missing translation for 'onlineSavingsMsg'
Learn More

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

48.74 € - 139.70 €

Chemical Identifiers

CAS 12009-21-1
Molecular Formula BaO3Zr
Molecular Weight (g/mol) 276.548
MDL Number MFCD00014189
InChI Key DQBAOWPVHRWLJC-UHFFFAOYSA-N
Synonym barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
PubChem CID 16212444
IUPAC Name barium(2+);dioxido(oxo)zirconium
SMILES [O-][Zr](=O)[O-].[Ba+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371559
SDS View Documents
Thermo Scientific Alfa Aesar
014028.30
250 g
48.74 €
250g
Please to purchase this item. Need a web account? Register with us today!
11361559
SDS View Documents
Thermo Scientific Alfa Aesar
014028.A1
1 kg
139.70 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.