missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium thiosulfate, pure, Thermo Scientific™
30.60 € - 431.00 €
Chemical Identifiers
CAS | 35112-53-9 |
---|---|
Molecular Formula | BaO3S2 |
Molecular Weight (g/mol) | 249.46 |
InChI Key | ONPIOWQPHWNPOQ-UHFFFAOYSA-L |
Synonym | barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo |
PubChem CID | 169663 |
IUPAC Name | barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
SMILES | [O-]S(=O)(=S)[O-].[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15229856
|
Thermo Scientific Acros
453811000 |
100 g |
30.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15239856
|
Thermo Scientific Acros
453815000 |
500 g |
101.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15219856
|
Thermo Scientific Acros
453810025 |
2.5 kg | |||||||
Description
Chemical Identifiers
35112-53-9 | |
249.46 | |
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
BaO3S2 | |
ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
169663 | |
[O-]S(=O)(=S)[O-].[Ba+2] |
Specifications
35112-53-9 | |
BaS2O3 | |
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
169663 | |
97.5 to 102.5% | |
Barium thiosulfate |
BaO3S2 | |
15,995 | |
Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol | |
[O-]S(=O)(=S)[O-].[Ba+2] | |
249.46 | |
249.46 | |
Pure |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 252-375-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Barium thiosulfate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only