Learn More
Barium thiosulfate, 98%, Thermo Scientific Chemicals
14.60 € - 126.00 €
Chemical Identifiers
CAS | 35112-53-9 |
---|---|
Molecular Formula | BaO3S2 |
Molecular Weight (g/mol) | 249.444 |
MDL Number | MFCD00040661 |
InChI Key | ONPIOWQPHWNPOQ-UHFFFAOYSA-L |
Synonym | barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo |
PubChem CID | 169663 |
IUPAC Name | barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
SMILES | [O-]S(=O)(=S)[O-].[Ba+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35112-53-9 | |
249.444 | |
ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
169663 | |
[O-]S(=O)(=S)[O-].[Ba+2] |
BaO3S2 | |
MFCD00040661 | |
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
Specifications
35112-53-9 | |
BaS2O3 | |
UN1564 | |
ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
169663 | |
98% | |
Barium thiosulfate |
BaO3S2 | |
MFCD00040661 | |
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
[O-]S(=O)(=S)[O-].[Ba+2] | |
249.444 | |
249.46 | |
3.45 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 252-375-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only