missing translation for 'onlineSavingsMsg'
Learn More

Barium selenite, 97%, Thermo Scientific™

Product Code. 11491990
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-59-7
Molecular Formula BaO3Se
Molecular Weight (g/mol) 264.295
MDL Number MFCD00003446
InChI Key VQKDPABSEYUERS-UHFFFAOYSA-L
Synonym barium selenite, selenious acid, barium salt 1:1, barium 2+ selenite, bariumselenit, barium 2+ selenit, acmc-20alqt, bariumselenite, ba.so3e, selenious acid barium salt, seleniousacid,bariumsalt 1:1
PubChem CID 166893
IUPAC Name barium(2+);selenite
SMILES [O-][Se](=O)[O-].[Ba+2]

Specifications

Quantity 500 g
Linear Formula BaSeO3
UN Number UN2630
Formula Weight 264.29
Percent Purity 97%
Chemical Name or Material Barium selenite
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'