Learn More
Barium perchlorate, anhydrous, Thermo Scientific Chemicals
61.51 € - 169.85 €
Chemical Identifiers
CAS | 13465-95-7 |
---|---|
Molecular Formula | BaCl2O8 |
Molecular Weight (g/mol) | 336.219 |
MDL Number | MFCD00003440 |
InChI Key | OOULUYZFLXDWDQ-UHFFFAOYSA-L |
Synonym | barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv |
PubChem CID | 61623 |
IUPAC Name | barium(2+);diperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361768
|
Thermo Scientific Alfa Aesar
011657.22 |
100 g |
61.51 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371768
|
Thermo Scientific Alfa Aesar
011657.36 |
500 g |
169.85 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13465-95-7 | |
336.219 | |
OOULUYZFLXDWDQ-UHFFFAOYSA-L | |
61623 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
BaCl2O8 | |
MFCD00003440 | |
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv | |
barium(2+);diperchlorate |
Specifications
13465-95-7 | |
Powder | |
MFCD00003440 | |
Soluble in water. | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] | |
336.219 | |
336.24 | |
Hygroscopic | |
Barium perchlorate |
Colorless | |
BaCl2O8 | |
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv | |
OOULUYZFLXDWDQ-UHFFFAOYSA-L | |
barium(2+);diperchlorate | |
61623 | |
Odorless | |
3.20 g/cm3 |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 236-710-4
missing translation for 'rtecsNumber' : SC7550000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.