missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Barium niobium oxide is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Powder |
Quantity | 50 g |
UN Number | UN1564 |
Solubility Information | Insoluble in water |
Formula Weight | 419.11 |
Percent Purity | 99.9% |
Assay | (metals basis) |
Chemical Name or Material | Barium niobium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'