Learn More
Barium metaborate monohydrate, Thermo Scientific™
18.00 € - 144.00 €
Chemical Identifiers
CAS | 19004-06-9 |
---|---|
Molecular Formula | B2BaH2O5 |
Molecular Weight (g/mol) | 240.96 |
MDL Number | MFCD00046182 |
InChI Key | JXOXZJCHHKIXMI-UHFFFAOYSA-N |
Synonym | barium metaborate monohydrate, 2bo2.ba.h2o, barium 2+ ;oxido oxo borane;hydrate, barium 2+ hydrate dioxoborinate, barium 2+ ion hydrate dioxoborinate |
PubChem CID | 23165673 |
SMILES | O.[Ba++].[O-]B=O.[O-]B=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19004-06-9 | |
240.96 | |
JXOXZJCHHKIXMI-UHFFFAOYSA-N | |
23165673 |
B2BaH2O5 | |
MFCD00046182 | |
barium metaborate monohydrate, 2bo2.ba.h2o, barium 2+ ;oxido oxo borane;hydrate, barium 2+ hydrate dioxoborinate, barium 2+ ion hydrate dioxoborinate | |
O.[Ba++].[O-]B=O.[O-]B=O |
Specifications
>900°C | |
White | |
B2BaH2O5 | |
MFCD00046182 | |
barium metaborate monohydrate, 2bo2.ba.h2o, barium 2+ ;oxido oxo borane;hydrate, barium 2+ hydrate dioxoborinate, barium 2+ ion hydrate dioxoborinate | |
O.[Ba++].[O-]B=O.[O-]B=O | |
240.96 | |
240.97 (222.96 Anhydrous) | |
Barium metaborate monohydrate |
19004-06-9 | |
Crystalline | |
BaB2O4·H2O | |
UN1564 | |
JXOXZJCHHKIXMI-UHFFFAOYSA-N | |
barium(2+) hydrate dioxoborinate | |
23165673 | |
3.35g/mL |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'tsca' : TSCA
Your input is important to us. Please complete this form to provide feedback related to the content on this product.