Learn More
Barium manganate(VI), 95%, Thermo Scientific Chemicals
18.28 € - 180.00 €
Chemical Identifiers
CAS | 7787-35-1 |
---|---|
Molecular Formula | BaMnO4 |
Molecular Weight (g/mol) | 256.26 |
MDL Number | MFCD00011514 |
InChI Key | ZZCNKSMCIZCVDR-UHFFFAOYSA-N |
Synonym | barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese |
PubChem CID | 3084030 |
IUPAC Name | barium(2+);dioxido(dioxo)manganese |
SMILES | [Ba++].[O-][Mn]([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10741345
|
Thermo Scientific Acros
198480050 |
5 g | Glass bottle | N/A | N/A | N/A | |||||
10348480
|
Thermo Scientific Acros
198480250 |
25 g | Glass bottle | N/A | N/A | N/A | |||||
10422011
|
Thermo Scientific Acros
198481000 |
100 g | Glass bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-35-1 | |
256.26 | |
ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
3084030 | |
[Ba++].[O-][Mn]([O-])(=O)=O |
BaMnO4 | |
MFCD00011514 | |
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
barium(2+);dioxido(dioxo)manganese |
Specifications
7787-35-1 | |
Fine Crystalline Powder | |
BaMnO4 | |
MFCD00011514 | |
15, 977 | |
Solubility in water: slightly soluble. Other solubilities: soluble in acids | |
[Ba++].[O-][Mn]([O-])(=O)=O | |
256.26 | |
256.27 | |
Barium manganate(VI), 95% |
Black-Blue to Blue | |
94% min | |
BaMnO4 | |
08,21; 09,23; 10,16; 12,38; 15,18; 16,18 | |
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
barium(2+);dioxido(dioxo)manganese | |
3084030 | |
95% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-109-6
missing translation for 'tsca' : TSCA
- Barium manganate(VI)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Oxidising solid Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.