missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥98%
51.07 € - 290.60 €
Chemical Identifiers
CAS | 12230-71-6 |
---|---|
Molecular Formula | BaH18O10 |
Molecular Weight (g/mol) | 315.46 |
MDL Number | MFCD00149152 |
InChI Key | ZUDYPQRUOYEARG-UHFFFAOYSA-L |
Synonym | barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate |
PubChem CID | 17749109 |
IUPAC Name | barium(2+);dihydroxide;octahydrate |
SMILES | O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15613260
|
Honeywell-Fluka
31127-100G |
100 g |
51.07 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15633260
|
Honeywell-Fluka
31127-500G |
500 g | N/A | ||||||
15623260
|
Honeywell-Fluka
31127-1KG |
1 kg |
290.60 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
barium(2+);dihydroxide;octahydrate |
Specifications
12230-71-6 | |
Ba(OH)2 · 8H2O | |
3262 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+);dihydroxide;octahydrate | |
17749109 | |
≥98% | |
Plastic Bottle |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.46g/mol | |
puriss. p.a. | |
Barium Hydroxide Octahydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302 + H332-H314
missing translation for 'einecsNumber' : 241-234-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Barium hydroxide octahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Barium hydroxide-8-hydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title