missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Monohydrate, purum p.a., ≥98.0% (T), Honeywell™

Product Code. 15631080
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]

Specifications

Boiling Point 780°C
Color White
pH 13.1 (21.0g/L at 25°C)
Quantity 1 kg
Linear Formula Ba(OH)2 · H2O
UN Number UN3262
Formula Weight 189.36g/mol
Percent Purity ≥98.0% (T)
Grade purum p.a.
Packaging Poly Bottle
Appearance Powder
Vapor Pressure 300 hPa at 78°C
Density 3.743g/mL at 25°C
Chemical Name or Material Barium Hydroxide Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'