Learn More
Barium dodecairon nonadecaoxide, Thermo Scientific Chemicals
88.40 € - 308.00 €
Chemical Identifiers
CAS | 12047-11-9 |
---|---|
Molecular Formula | BaFe12O19 |
MDL Number | MFCD00075637 |
Synonym | Barium ferrite |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15422357
|
Thermo Scientific Alfa Aesar
045552.14 |
25 g |
88.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432357
|
Thermo Scientific Alfa Aesar
045552.22 |
100 g |
308.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium dodecairon nonadecaoxide is used in subwoofer magnets to provide power. It is also used in new high tech magnetic tapes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12047-11-9 | |
BaFe12O19 | |
MFCD00075637 | |
Barium ferrite |
Specifications
>1315°C | |
Black | |
BaFe12O19 | |
UN1564 | |
Insoluble in water. | |
Barium dodecairon nonadecaoxide |
12047-11-9 | |
Powder | |
MFCD00075637 | |
Barium ferrite | |
1111.48 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 234-974-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only