Learn More
Barium copper oxide (1-1), 99.9% (metals basis), Thermo Scientific™
Preparation of superconducting powders
74.00 € - 296.00 €
Specifications
Chemical Name or Material | Barium copper oxide (1-1) |
---|---|
Physical Form | Fine Agglomerate |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00799047 |
UN Number | UN1564 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Barium copper oxide (1-1) | |
(metals basis) | |
UN1564 |
Fine Agglomerate | |
MFCD00799047 | |
99.9% |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Hazard Class: 6.1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.