missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, ≥99%, Honeywell Fluka™

Product Code. 15677690
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 962°C
Color Colorless
pH 0k
Physical Form Solid
Quantity 1 kg
UN Number UN1564
IUPAC Name barium(2+);dichloride;dihydrate
Formula Weight 244.26g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly Bottle
Density 3.100g/cm3
Chemical Name or Material Barium chloride dihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'