Learn More
Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
242.00 € - 2271.00 €
Chemical Identifiers
CAS | 513-77-9 |
---|---|
Molecular Formula | CBaO3 |
Molecular Weight (g/mol) | 197.34 |
MDL Number | MFCD00003448 |
InChI Key | AYJRCSIUFZENHW-UHFFFAOYSA-L |
Synonym | barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p |
PubChem CID | 10563 |
SMILES | [Ba++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300849
|
Thermo Scientific Alfa Aesar
010645.14 |
25 g | |||||||
11380839
|
Thermo Scientific Alfa Aesar
010645.22 |
100 g |
711.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390839
|
Thermo Scientific Alfa Aesar
010645.36 |
500 g | |||||||
Description
Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-77-9 | |
197.34 | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
10563 |
CBaO3 | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
[Ba++].[O-]C([O-])=O |
Specifications
(decomposition) | |
513-77-9 | |
CBaO3 | |
14,969 | |
Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid. | |
[Ba++].[O-]C([O-])=O | |
197.34 | |
197.35 | |
Puratronic™ | |
4.43 g/mL | |
Barium carbonate |
811°C | |
Lumps, Flakes and Granular Powder | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
barium(2+) carbonate | |
10563 | |
99.997% | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 208-167-3
missing translation for 'rtecsNumber' : CQ8600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only