missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99%, Thermo Scientific Chemicals

 

31.96 € - 331.65 €

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374599
SDS View Documents
Thermo Scientific Alfa Aesar
A13195.22
100 g
31.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
11384599
SDS View Documents
Thermo Scientific Alfa Aesar
A13195.36
500 g
49.75 €
500g
Please to purchase this item. Need a web account? Register with us today!
11424614
SDS View Documents
Thermo Scientific Alfa Aesar
A13195.0B
1 kg
79.80 €
1000g
Please to purchase this item. Need a web account? Register with us today!
11434614
SDS View Documents
Thermo Scientific Alfa Aesar
A13195.0I
5 kg
331.65 €
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.

Solubility
Insoluble in water and alcohol. Soluble in acetic acid, hydrochloric acid and nitric acid.

Notes
m.p. 174° (90 atm), 811° (1 atm)
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 208-167-3

missing translation for 'rtecsNumber' : CQ8600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium carbonate, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.