missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals

59.50 € - 153.77 €

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326598
SDS View Documents
Thermo Scientific Alfa Aesar
012197.22
100 g
59.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
11336598
SDS View Documents
Thermo Scientific Alfa Aesar
012197.36
500 g
153.77 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used  precursor to chemicals used in photograph.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-28-8
333.17
HQQMRSGBIPFNSN-UHFFFAOYSA-L
O.O.[Br-].[Br-].[Ba++]
BaBr2H4O2
MFCD00149153
barium(2+) dihydrate dibromide

Specifications

7791-28-8
99.30%
MFCD00149153
HQQMRSGBIPFNSN-UHFFFAOYSA-L
barium(2+) dihydrate dibromide
333.17 (297.14 Anhydrous)
Barium bromide dihydrate
Crystalline
BaBr2H4O2
Very soluble in water,soluble in methanol
O.O.[Br-].[Br-].[Ba++]
333.17
3.58
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 234-140-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.