Learn More
Barium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
For ultra trace analysis
76.58 € - 259.29 €
Chemical Identifiers
CAS | 10553-31-8 |
---|---|
Molecular Formula | BaBr2 |
Molecular Weight (g/mol) | 297.16 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15229826
|
Thermo Scientific Acros
315380500 |
50 g |
76.58 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15239826
|
Thermo Scientific Acros
315382500 |
250 g |
259.29 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10553-31-8 | |
BaBr2 | |
297.16 |
Specifications
850.0°C | |
98.5% min. (Argentometry) | |
BaBr2 | |
Solubility in water: 104.1g/100mL (20°C). Other solubilities: soluble in methanol, almost insoluble in ethanol, ethyl acetate,, acetone, dioxane | |
297.16 | |
Pure |
10553-31-8 | |
BaBr2 | |
15,964 | |
297.16 | |
99% | |
Barium bromide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 234-140-
- Barium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Barium bromide
RUO – Research Use Only