Learn More
Barium acetate, 99+%, ACS reagent, Thermo Scientific Chemicals
25.43 € - 243.21 €
Chemical Identifiers
CAS | 543-80-6 |
---|---|
Molecular Formula | C4H6BaO4 |
Molecular Weight (g/mol) | 255.42 |
MDL Number | MFCD00012447 |
InChI Key | ITHZDDVSAWDQPZ-UHFFFAOYSA-L |
Synonym | barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate |
PubChem CID | 10980 |
IUPAC Name | barium(2+);diacetate |
SMILES | [Ba++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10583494
|
Thermo Scientific Acros
423431000 |
100 g | Plastic bottle |
25.43 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10197890
|
Thermo Scientific Acros
423435000 |
500 g | Plastic bottle |
73.87 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10608994
|
Thermo Scientific Acros
423430025 |
2.5 kg | Plastic bottle |
243.21 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
543-80-6 | |
255.42 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
10980 | |
[Ba++].CC([O-])=O.CC([O-])=O |
C4H6BaO4 | |
MFCD00012447 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
barium(2+);diacetate |
Specifications
543-80-6 | |
Crystals | |
C4H6BaO4 | |
MFCD00012447 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
barium(2+);diacetate | |
10980 | |
≥99% | |
2.46 | |
Barium acetate |
White | |
99.0 to 102% (Complexometry) | |
(CH3CO2)2Ba | |
15, 961 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
[Ba++].CC([O-])=O.CC([O-])=O | |
255.42 | |
255.43 | |
ACS Reagent | |
2.4600g/mL |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 208-849-0
missing translation for 'rtecsNumber' : AF4550000
missing translation for 'tsca' : TSCA
- Barium acetate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.