Learn More
Masterflex™ Y-Connector, Barbed, Polypropylene
Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.
Brand:  Masterflex™ 06296-10
 View more versions of this product
Product Code. 12459580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.
  • Ideal for the following:
    • Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Designed for quick and easy assembly with flexible tubing
  • Available in a variety of barb sizes, fitting lengths, and fitting widths
Specifications
Specifications
Y-shaped, Barbed
4-6mm ID Tubing
10/Pk.
4-6mm ID
Autoclave
Y
White
32 mm
Low pressure applications. Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Tubing connector
6mm, 6mm, 6mm
36 mm
Polypropylene
30 psi (2 bar)