missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Y-Connector, Barbed, Polypropylene

Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.

Brand:  Masterflex™ 06295-70

 View more versions of this product

Product Code. 12439590

  • 55.20 € / Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.

  • Ideal for the following:
    • Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Designed for quick and easy assembly with flexible tubing
  • Available in a variety of barb sizes, fitting lengths, and fitting widths
Specifications

Specifications

Y-shaped, Barbed
96 mm
Low pressure applications. Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Tubing connector
16mm, 16mm, 16mm
61 mm
Clear
Polypropylene
16mm ID Tubing
10/Pk.
16mm ID
Autoclave
Y
30 psi (2 bar)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers