missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Y-Connector, Barbed, Polypropylene

Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.

Brand:  Masterflex™ 06295-10

 View more versions of this product

Product Code. 11795888

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Cole-Parmer™ Y-Connector, Barbed, Polypropylene is a simple, economical fitting to be used with flexible tubing in low pressure applications.

  • Ideal for the following:
    • Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Designed for quick and easy assembly with flexible tubing
  • Available in a variety of barb sizes, fitting lengths, and fitting widths
Spezifikation

Spezifikation

Y-shaped, Barbed
24 mm
Low pressure applications. Pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Tubing connector
4mm, 4mm, 4mm
14 mm
Clear
Polypropylene
4mm ID Tubing
20/Pk.
4mm ID
Autoclave
Y
30 psi (2 bar)
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen
Sonderangebote

Sonderangebote