Learn More
Azodicarbonamide, 97%, Thermo Scientific Chemicals
40.50 €
Chemical Identifiers
CAS | 123-77-3 |
---|---|
Molecular Weight (g/mol) | 116.08 |
MDL Number | MFCD00007958 |
InChI Key | XOZUGNYVDXMRKW-AATRIKPKSA-N |
Synonym | azodicarbonamide, azodicarboxamide, diazenedicarboxamide, azobisformamide, azodiformamide, azodicarbamide, azodicarboamide, azobiscarbonamide, azobiscarboxamide, celogen az |
PubChem CID | 5462814 |
IUPAC Name | (E)-carbamoyliminourea |
SMILES | C(=O)(N)N=NC(=O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10589620
|
Thermo Scientific Acros
105165000 |
500 g | Plastic bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen)Chemical Identifiers
123-77-3 | |
MFCD00007958 | |
azodicarbonamide, azodicarboxamide, diazenedicarboxamide, azobisformamide, azodiformamide, azodicarbamide, azodicarboamide, azobiscarbonamide, azobiscarboxamide, celogen az | |
(E)-carbamoyliminourea |
116.08 | |
XOZUGNYVDXMRKW-AATRIKPKSA-N | |
5462814 | |
C(=O)(N)N=NC(=O)N |
Specifications
123-77-3 | |
97% | |
H2NCON=NCONH2 | |
03,IV,246 | |
azodicarbonamide, azodicarboxamide, diazenedicarboxamide, azobisformamide, azodiformamide, azodicarbamide, azodicarboamide, azobiscarbonamide, azobiscarboxamide, celogen az | |
XOZUGNYVDXMRKW-AATRIKPKSA-N | |
(E)-carbamoyliminourea | |
5462814 | |
97% |
195.0°C to 202.0°C | |
C2H4N4O2 | |
MFCD00007958 | |
15,912 | |
Solubility in water: soluble in hot water. Other solubilities: insoluble in common solvents, soluble in dimethyl sulfoxide, slightly soluble in demethylformamide | |
C(=O)(N)N=NC(=O)N | |
116.08 | |
116.08 | |
Azodicarbonamide, 97% |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-650-8
missing translation for 'rtecsNumber' : LQ1040000
missing translation for 'tsca' : TSCA
- Azodicarbonamide
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.