missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Tapered Glass Joint Sleeve
Forms a liquid and gas tight seal.
Brand: Azlon SWN802
This item is not returnable.
View return policy
Description
- For grease free and high vacuum seals using Quickfit™ ground glass joints
- Strong construction with knurled shoulder for ease of handling and repeated use
- Self lubricating and non-contaminating
- Working temperature range -100°C to +260°C
- Working vacuum ≤10-3atmospheres
- Highly resistant to chemical attack
Specifications
PTFE | |
1/Pk. | |
10/19 |
Tapered sleeves | |
−100 to 250°C |