missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polyethylene Cylindrical Pneumatic Trough

Cylindrical, one piece pneumatic troughs

Brand:  Azlon TGJ062

 View more versions of this product

Product Code. 12018943

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Much lighter than glass
  • Will withstand up to 80°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flat
Cylindrical
150 mm
Polyethylene
Pneumatic Trough
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers