Learn More
Azetidine-3-carboxylic acid, 98+%, Thermo Scientific Chemicals
42.60 € - 451.00 €
Chemical Identifiers
CAS | 36476-78-5 |
---|---|
Molecular Formula | C4H7NO2 |
Molecular Weight (g/mol) | 101.11 |
MDL Number | MFCD00191763 |
InChI Key | GFZWHAAOIVMHOI-UHFFFAOYSA-N |
Synonym | 3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid |
PubChem CID | 93192 |
IUPAC Name | azetidine-3-carboxylic acid |
SMILES | OC(=O)C1CNC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488917
|
Thermo Scientific Alfa Aesar
H52294.MD |
250 mg |
42.60 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15468917
|
Thermo Scientific Alfa Aesar
H52294.03 |
1 g |
126.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478917
|
Thermo Scientific Alfa Aesar
H52294.06 |
5 g |
451.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
36476-78-5 | |
101.11 | |
GFZWHAAOIVMHOI-UHFFFAOYSA-N | |
93192 | |
OC(=O)C1CNC1 |
C4H7NO2 | |
MFCD00191763 | |
3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid | |
azetidine-3-carboxylic acid |
Specifications
36476-78-5 | |
C4H7NO2 | |
3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid | |
GFZWHAAOIVMHOI-UHFFFAOYSA-N | |
azetidine-3-carboxylic acid | |
93192 | |
≥98% |
∽250°C (decomposition) | |
MFCD00191763 | |
Soluble in water. | |
OC(=O)C1CNC1 | |
101.11 | |
101.11 | |
Azetidine-3-carboxylic acid |
Safety and Handling
missing translation for 'rtecsNumber' : CM4310600
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only