Learn More
Axygen™ Axypet™ 8-Channel Digital Pipettor, Calibration 3x4 ALT
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
710.00 €
Specifications
Certifications/Compliance ISO17025
Number of Channels 8
Product Line Axypet™
Type Multi-channel Pipettor
Quantity 1/Pk., 1/Cs
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17213634
View Documents
Axygen™
AP-8-10-ALT
0.5 to 10 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17273634
View Documents
Axygen™
AP-8-50-ALT
5 to 50 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17233634
View Documents
Axygen™
AP-8-200-ALT
20 to 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17253634
View Documents
Axygen™
AP-8-300-ALT
50 to 300 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
  • Push button and ejector button are ergonomically located
  • Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
  • Tip manifold rotates 360°
  • Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
  • Shafts withdraw when pressed against a row of tips
  • Each shaft moves independently, minimizing jamming of tips
  • Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifications
Specifications
ISO17025
Axypet™
1/Pk., 1/Cs
8
Multi-channel Pipettor