Learn More
Axygen™ Macrovolume Pipet Tips, For Axypet-16
For use with Axypet-16 pipettes
Brand: Axygen™ T-1536-R
This item is not returnable.
View return policy
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery™, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Maxymum Recovery technology for ultra-low retention
Specifications
0.5 to 30 μL | |
Clear | |
DNase, RNase, ATP, and Endotoxin Free | |
Microvolume | |
Axypet™-16 |
Non-sterile | |
Autoclavable | |
Rack | |
384 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polypropylene |