Learn More
Axygen™ 96-well Low Profile Bar Coded PCR Microplates
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The low profile design results in faster and more efficient PCR reactions.
Brand: Axygen™ PCR-96-LP-AB-C-PBC
This item is not returnable.
View return policy
Description
- Low profile; Optimized for fast PCR
- Certified RNase-/DNase-free
- Nonpyrogenic, nonsterile
- Bar coded for traceability and identification
- Sealing options: Axymats, heat sealing, adhesive films and foils, flat and domed cap strips
Specifications
Includes Barcode | |
Standard | |
Clear | |
Polypropylene | |
Non-sterile |
RNase-/DNase-free | |
8 x 12 | |
ABI thermal cyclers, ABI Fast PCR 7500, 7900 9800, Veriti and StepOne | |
96 | |
PCR thermal transfer, RT-PCR applications |