Learn More
Axygen™ 1200μL Universal Pipetter Tips
Fits most popular brands of pipettes
Brand: Axygen™ T-1200-C
This item is not returnable.
View return policy
Description
- Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
- Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
- Volume range: 100 to 1200μL
- Available in bulk or racked formats
Specifications
1200 μL | |
White | |
Genomic/Wide Orifice, Universal | |
Universal Fit |