Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Fixed Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand:  Trajan Scientific and Medical™ 0029841
 View more versions of this product
Product Code. 15248463
  • 102.00 € / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
CTC/Thermo (Classic Button) Syringe
0.63 mm
10 μL
Thermo Gas Chromatography
23 G
Removable Needle
Cone