missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Autophagy Inhibitor, 3-MA

66.03 € - 112.00 €

Chemical Identifiers

CAS 5142-23-4
Molecular Formula C6H7N5, C6H7N5
Synonym 3-Methyladenine; 6-Amino-3-methylpurine
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15406599
View Documents
Thermo Scientific Chemicals
J64813.MB
25 mg
66.03 €
25mg
Please to purchase this item. Need a web account? Register with us today!
15416599
View Documents
Thermo Scientific Chemicals
J64813.MF
50 mg
112.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
Specifications

Chemical Identifiers

5142-23-4
C6H7N5, C6H7N5
3-Methyladenine; 6-Amino-3-methylpurine

Specifications

White
A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
C6H7N5, C6H7N5
3-Methyladenine; 6-Amino-3-methylpurine
Solid
146087
CN1C=NC(N)=C2N=CN=C12
149.16
2°C to 8°C
5142-23-4
149.15g/mol
Signal Transduction Reagent
MFCD00010531
FSASIHFSFGAIJM-UHFFFAOYSA-N
3-methyl-3H-purin-6-amine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 225-908-6

Recommended Storage : Keep cold

Product Identifier
  • Autophagy Inhibitor, 3-MA

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

Supplemental information
  • MIXTURE LIST-Contains : 3-Methyl-3H-adenine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.