Learn More
Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System
GREENER_CHOICE
Increase pipetting performance through superior tip straightness and low %CV with these pipette tips for the Molecular Devices system.
Brand:  Thermo Scientific™ 937-261
 View more versions of this product
Product Code. 15252999
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Molecular Device FLIPR with Type A CCS 384-well head and Type B Liberty 384-well head.
  • Available in non-sterile, sterile and with the Thermo Scientific™ ART™ filter technology
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 μL
Sterile
Rack
Molecular Devices FLIPR Type A CCS 384-well Head
Non-filtered
Clear
Automation
Molecular Devices
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.