Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand: Thermo Scientific™ 985-021
This item is not returnable.
View return policy
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
- Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifications
80 μL | |
Sterile | |
Rack | |
BioTek Pipetting System |
Filtered | |
Clear | |
Low Retention | |
BioTek |