missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Crystalline Powder, 20 to 200 Mesh, U.S.P. - F.C.C., J.T. Baker™
373.00 € - 713.00 €
Chemical Identifiers
CAS | 50-81-7 |
---|---|
Molecular Formula | C6H8O6 |
Molecular Weight (g/mol) | 176.12 |
MDL Number | MFCD00064328 |
InChI Key | CIWBSHSKHKDKBQ-DOMZIZNONA-N |
Synonym | l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin |
PubChem CID | 54670067 |
ChEBI | CHEBI:29073 |
IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
SMILES | OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15557874
|
Avantor J.T.Baker
0936-07 |
500 g | Poly Bottle |
373.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15567874
|
Avantor J.T.Baker
0936-08 |
1 kg | Poly Bottle |
713.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
50-81-7 | |
176.12 | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
54670067 | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
C6H8O6 | |
MFCD00064328 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
CHEBI:29073 | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Specifications
50-81-7 | |
C6H8O6 | |
1L = 1.65kg | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
54670067 | |
176.12 | |
Crystalline Powder |
20 to 200 Mesh | |
MFCD00064328 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
176.12 | |
CHEBI:29073 | |
FCC/USP | |
Ascorbic Acid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title