Learn More
Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, 10, bulk
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Brand:  Thermo Scientific™ 2140B
Product Code. 10647351
Description
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ART 10 REACH, Filtered, Non-sterile, Bulk
Filtered
ART 10 REACH
4.33cm
Extended Length
ART
10 μL
Non-sterile
1.71 in.
Bulk
Universal Fit