missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Arsenic(V) oxide, 99.9% (metals basis)

135.68 € - 421.10 €

Chemical Identifiers

CAS 1303-28-2
Molecular Formula As2O5
Molecular Weight (g/mol) 229.838
MDL Number MFCD00003434
InChI Key COHDHYZHOPQOFD-UHFFFAOYSA-N
Synonym arsenic pentoxide, arsenic v oxide, arsenic anhydride, arsenic v pentoxide, arsenic pentaoxide, arsenic oxide, diarsenic pentoxide, diarsenic pentaoxide, arsenic 5+ oxide, arsenic acid anhydride
PubChem CID 14771
SMILES O=[As](=O)O[As](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326978
SDS View Documents
Thermo Scientific Chemicals
014668.14
25 g
135.68 €
25g
Please to purchase this item. Need a web account? Register with us today!
11336978
SDS View Documents
Thermo Scientific Chemicals
014668.22
100 g
421.10 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Arsenic(V) oxide is used in the preparation of metal arsenates. It is used for dye and printing industries. Further, it is used in wood preservatives, colored glass and metal adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-28-2
229.838
COHDHYZHOPQOFD-UHFFFAOYSA-N
14771
As2O5
MFCD00003434
arsenic pentoxide, arsenic v oxide, arsenic anhydride, arsenic v pentoxide, arsenic pentaoxide, arsenic oxide, diarsenic pentoxide, diarsenic pentaoxide, arsenic 5+ oxide, arsenic acid anhydride
O=[As](=O)O[As](=O)=O

Specifications

315°C (decomposition)
White
99.9% (metals basis)
MFCD00003434
14,798
Soluble in water and alcohol.
O=[As](=O)O[As](=O)=O
14771
99.9%
Hygroscopic
(metals basis)
1303-28-2
Powder
As2O5
UN1559
arsenic pentoxide, arsenic v oxide, arsenic anhydride, arsenic v pentoxide, arsenic pentaoxide, arsenic oxide, diarsenic pentoxide, diarsenic pentaoxide, arsenic 5+ oxide, arsenic acid anhydride
COHDHYZHOPQOFD-UHFFFAOYSA-N
229.838
229.79
Odorless
4.32 g/mL
Arsenic(V) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: ARSENIC PENTOXIDE

missing translation for 'einecsNumber' : 215-116-9

missing translation for 'rtecsNumber' : CG2275000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Arsenic(V) oxide, 99.9% (metals basis)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.