missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11378848
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]

Specifications

Melting Point 381°C
Physical Form Pieces
Quantity 50 g
UN Number UN1557
Formula Weight 532.64
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.5
Density 6.5
Chemical Name or Material Arsenic(III) telluride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'