missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals

78.29 € - 195.98 €

Chemical Identifiers

CAS 12044-79-0
Molecular Formula AsH3S
Molecular Weight (g/mol) 110.01
MDL Number MFCD00049841
InChI Key BMOFEAKQXGGUPE-UHFFFAOYSA-N
Synonym arsenic ii sulfide, realgar ass, diarsanedithione
IUPAC Name arsanethiol
SMILES S[AsH2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323337
SDS View Documents
Thermo Scientific Alfa Aesar
017284.06
5 g
78.29 €
5g
Please to purchase this item. Need a web account? Register with us today!
11333337
SDS View Documents
Thermo Scientific Alfa Aesar
017284.14
25 g
195.98 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Arsenic(II) sulfide is used as an primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.