missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARNTL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14268-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
ARNTL | |
Polyclonal | |
Unconjugated | |
ARNTL | |
ARNT3, ARNTL, bHLH PAS protein JAP3, bHLHe5, BMAL1, BMAL1c, Brain and muscle ARNT like 1, JAP3, Member of PAS protein 3, MOP3, PASD3, TIC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11865, 29657, 406 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00327, Q9EPW1, Q9WTL8 | |
ARNTL | |
ARNTL Fusion Protein Ag5586 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |