Learn More
Arginase-1 Mouse anti-Human, CoraLite 488, Clone: 5D6D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66129-100UL
This item is not returnable.
View return policy
Description
The Arginase-1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Arginase-1. This antibody recognizes human, pig, rat, mouse antigen. The Arginase-1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Arginase-1 | |
Monoclonal | |
1670 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Immunofluorescence | |
5D6D12 | |
CoraLite 488 | |
100 μL | |
383 | |
IgG1 |